Learn More
Zeiss™ Stemi™ 2000/2000C Stereomicroscope Auxiliary Objectives
Use this objective with Cal Zeiss Mocroscopes.
Brand:  Zeiss™ 455026-0000-000
Product Code. 11876472
  • 361.00€ / Each
Only null left
Estimated Shipment: 04-08-2025
Description
  • Front lens system
  • 0.4x
  • FWD 211mm
Specifications
Specifications
Lens System
Stereomicroscope
Stemi™ 2000, 2000C
0.4X