Learn More
C2orf37 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26033-1-AP-20UL
This item is not returnable.
View return policy
Specifications
C2orf37 | |
Polyclonal | |
Unconjugated | |
DCAF17 | |
C2orf37, DCAF17 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
75763, 80067 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q3TUL7, Q5H9S7 | |
Dcaf17 | |
C2orf37 Fusion Protein Ag21995 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Product Title
C2orf37 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated