Learn More
C11orf80 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25957-1-AP-20UL
This item is not returnable.
View return policy
Description
C11orf80 locates at the chromosome 11. It has two isoforms with MW 70-75 kDa.Specifications
C11orf80 | |
Polyclonal | |
Unconjugated | |
TOP6BL | |
C11orf80 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79703 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8N6T0 | |
TOP6BL | |
C11orf80 Fusion Protein Ag22895 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |