Learn More
Buffer solution pH 7.00 (+/-0.024 @ 25oC) Colored Green Specpure NIST Traceable
Product Code. p-4781823
Click to view available options
Quantity:
500 mL
1 L
4 L
Unit Size:
1 L
4 L
500mL
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Chemical Name or Material Buffer solution
Name Note NIST Traceable
CAS 1336-21-6
Color Green
Physical Form Liquid
Quantity 500 mL
Molecular Formula H5NO
MDL Number MFCD00066650
Traceability to NIST Yes
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Solubility Information Miscible with water.
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name azanium;hydroxide
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Grade Specpure™