Learn More
Brand™ Counting Chambers, Burker-Turk pattern, BLAUBRAND™, double ruling
BLAUBRAND™ quality. Outstanding measurement precision. 100% certified quality.
Brand:  Brand™ 719520
975.09 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10297390
  • 1005.00 NOK / Each
Only null left
Estimated Shipment: 09-09-2025
Description
  • Manufactured according to DIN 12 847, including 2 haemacytometer cover glasses.
  • Combination of the Burker and Thoma systems.The ruling shows 9 large squares of 1 mm2 each.
  • The large squares are subdivided into 16 group squares with 0.2 mm sides. in the central large square, each group square is subdivided into 16 mini squares with 0.05 mm sides ( 0.0025 mm2).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Counting Device
Double-ruling, w/ Spring Clips
1Each
0.1mm