Learn More
BUCHI Calibration and Verification Standards for BUCHI Melting Point Instruments
Utilize the embedded firmware and software on your Melting Point apparatus to calibrate and verify your instrument at any time.
Brand: BUCHI 037454
View more versions of this product
Includes: Vanillin, Caffeine, Phenacetin
Description
Utilize the embedded firmware and software on your BUCHI Melting Point apparatus to calibrate and verify your instrument at any time.
- Comprehensive IQ/OQ procedures including certified calibration and verification compounds for GLP/GMP laboratories
- Guided calibrations and verifications provide easy and reliable results
- Ensures compliance from 10°C to 400°C
Specifications
Calibration Kit | |
BUCHI B-540/545 Melting Point apparatus | |
Vanillin, caffeine, phenacetin |