Learn More
Brannan™ BS 718 : 1991 Density Hydrometer L50
For reporting the measured specific gravity of a liquid relative to water. Brannan™ BS 718 : 1991 Density Hydrometer L50 is aligned with ISO 387 and ISO 649 and is manufactured from soda glass, steel shot ballast and synthetic wax resin.
Brand:  Brannan™ 10/363/0
 View more versions of this product
Product Code. 15727847
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • BS 718 : 1991 is generally aligned with ISO 387 and ISO 649
  • Calibrated using the accepted units of grams per milliliter (g/mL)
  • Available for numerous specific gravity ranges
  • Available for both low and medium surface tension
  • Includes protective plastic tube
  • 330mm in length
Specifications
Specifications
Density Hydrometer
0.0005 g/mL
330 mm
23 to 27 mm (Bulb)
103 g
For low surface tension
0.0005 g/mL
ISO 387 and ISO 649
Protective plastic tube (47 g)
Soda glass, Steel shot ballast, Synthetic wax resin
56 g
0.75 to 0.80 g/mL