Learn More
Bromocyclobutane, 95%, Thermo Scientific Chemicals
Product Code. 11465193
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
CAS: 4399-47-7 | C4H7Br | 135.004 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4399-47-7
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.004
MDL Number MFCD00001317
InChI Key KXVUSQIDCZRUKF-UHFFFAOYSA-N
Synonym cyclobutyl bromide, cyclobutane, bromo, cyclobutylbromid, cyclobutylbromide, bromo cyclobutane, bromo-cyclobutane, bromocyclobutane, acmc-209jw1, ksc235o4p, inchi=1/c4h7br/c5-4-2-1-3-4/h4h,1-3h
PubChem CID 78110
IUPAC Name bromocyclobutane
SMILES C1CC(C1)Br
Specifications
Density 1.431
Boiling Point 107°C to 108°C
Flash Point 22°C (71°F)
Refractive Index 1.4795
Quantity 5 g
UN Number UN1993
Beilstein 1900415
Sensitivity Light sensitive
Formula Weight 135.01
Percent Purity 95%
Chemical Name or Material Bromocyclobutane
Product Title
Bromocyclobutane, 95%, Thermo Scientific Chemicals >