Learn More
BRANDTECH™ VITLAB™ POM or PMP Forceps
POM or PMP forceps offer resilience and flexibility. Available in various lengths and tip styles.
179.00 NOK
Product Code | Brand | Length (Metric) | Tip Style | Material | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Tip Style | Material | Price | Quantity & Availability | |||
10377604
|
BRANDTECH™ VITLAB™
68299 |
180 mm | Blunt | POM | |||||
Description
POM (polyoxymethylene) forceps:
- Color: yellow
- External finger grooves for exceptional grip
- Blunt tips
- Part No. 68299 and 68399 have grooves inside of tip
- Part No. 68099 and 68199 are without grooves
PMP (polymethylpentene) forceps:
- Color: white
- Pointed tips
Specifications
Forceps | |
Standard |