Learn More
Mutexil™ Charlotte Disposable Round Caps
Charlotte caps are ideal in clinic, hospital, restaurant, and food processing settings. Eco-friendly, disposable caps are elastic and are constructed of non-woven polypropylene.
935.00 NOK - 1090.00 NOK
Specifications
Product Type | Bouffant |
---|---|
Material | Polypropylene |
Color | White |
Disposable | Single-use |
Product Code | Brand | Diameter (Metric) | Product Type | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Product Type | Quantity | Price | Quantity & Availability | |||
11795463
|
Mutexil™
4522090 |
53 cm | Bouffant | 1000/Pk. |
935.00 NOK
Pack of 1000 |
Estimated Shipment: 12-08-2025
Log in to see stock. |
|||
15315612
|
Mutexil™
4561325 |
48 cm | Bouffant | 1000/Pk. |
1090.00 NOK
Pack of 1000 |
N/A | |||
Description
- Round
- Polypropylene
- Non-woven
- White
Specifications
Bouffant | |
Polypropylene | |
White | |
Single-use |