Learn More
3M™ Blue SMS Based Fabric Category III Protective Coveralls
550.00 DKK
Specifications
Product Type | General Purpose Coveralls |
---|---|
Gender | Unisex |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Elastic |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11983764
|
3M™
7000089624 |
4X-Large | |||||||
Description
Specifications
General Purpose Coveralls | |
Single-use | |
Elastic | |
Hooded | |
Serged | |
Category III |
Unisex | |
Elastic | |
Elastic | |
2-panel | |
Non-sterile |