Learn More
OHAUS™ Block Cover
OHAUS™ Block Cover is a clear cover for use with low temperature blocks.
Brand: OHAUS™ 30400180
1438.99 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Block heater accessories include modular blocks to accommodate almost any sample
- Modular blocks feature close contact of tubes-to-walls allowing for maximum heat retention
- Most blocks feature a thermometer well for measuring block temperature
- Stainless steel sand baths are available for irregular samples when there is no suitable block available
- Low temperature covers help to reduce the air flow for additional temperature stability
Specifications
Cover Low-Temperature 4 Block | |
Accessory, Block Cover | |
Low Temperatures |