Learn More
BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flask with Polypropylene Stopper, with Certificate
Constructed of Borosilicate Glass and complies with DIN EN ISO 1042 standards. DE-M marking on the neck provides quick and accurate volume check. Polypropylene stopper included. Delivered with individual conformity certificate.
636.00 NOK - 2095.00 NOK
Specifications
Calibration | Calibrated To Contain |
---|---|
Certifications/Compliance | DIN EN ISO 1042 |
Closure Material | Polypropylene |
Color | Clear |
Description | DE-M Marking |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||
15805542
|
BRAND™
937243 |
10 mL | 10/19 |
636.00 NOK
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
||||
15825542
|
BRAND™
937246 |
20 mL | 10/19 |
641.00 NOK
Each |
Estimated Shipment: 30-09-2025
Log in to see stock. |
||||
15815542
|
BRAND™
937245 |
20 mL | 12/21 |
641.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15835542
|
BRAND™
937247 |
25 mL | 10/19 |
641.00 NOK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
||||
15895542
|
BRAND™
937290 |
50 mL | 14/23 |
656.00 NOK
Each |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||
15845542
|
BRAND™
937250 |
200 mL | 14/23 |
697.00 NOK
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
||||
15521291
|
BRAND™
937252 |
500 mL | 19/26 |
749.00 NOK
Each |
Estimated Shipment: 29-09-2025
Log in to see stock. |
||||
15855542
|
BRAND™
937253 |
1000 mL | 24/29 |
969.00 NOK
Each |
Estimated Shipment: 19-09-2025
Log in to see stock. |
||||
15865542
|
BRAND™
937254 |
2000 mL | 29/32 |
1215.00 NOK
Each |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||
15894968
|
BRAND™
937238 |
5 mL | 10/19 |
636.00 NOK
Each |
Estimated Shipment: 08-10-2025
Log in to see stock. |
||||
12304723
|
BRAND™
937255 |
5 L | 34/35 |
2095.00 NOK
Each |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||
Description
- Class A Volumetric Flask
- Constructed of Borosilicate Glass
- Complies with DIN EN ISO 1042 standards
- Delivered with individual conformity certificate
- Calibrated to contain (TC, In)
- Narrow neck and wide neck options available in a variety of sizes
- DE-M marking provides quick and accurate volume check
- Polypropylene stopper included
Specifications
Calibrated To Contain | |
Polypropylene | |
DE-M Marking | |
Yes | |
Stopper |
DIN EN ISO 1042 | |
Clear | |
Borosilicate Glass | |
A |