Learn More
Kautex™ Black Polypropylene Screw Closures
Brand: Kautex™ 2000971152
This item is not returnable.
View return policy
Description
Specifications
Black | |
Polypropylene | |
Polyethylene Foam Insert | |
Screw Cap | |
32 mm |
w/out Tamper Evident Ring | |
00 mm | |
00 mm | |
100 mL Bottle |