Learn More
Fisherbrand™ Black Phenolic PolyCone Cap
Forms an exceptionally tight seal and offer a good chemical barrier. Fisherbrand™ Black Phenolic PolyCone Caps are crack resistant.
2155.03 NOK - 11535.56 NOK
Specifications
Color | Black |
---|---|
Material | Phenolic Resin |
Liner Material | PolyCone |
Product Type | Polycone Cap |
Product Code | Brand | Neck Finish | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Neck Finish | Quantity | Price | Quantity & Availability | ||||
11538172
|
Fisherbrand™
02911803 |
20-400 | 5500/Cs. | ||||||
15592351
|
Fisherbrand™
02912055 |
24-400 | 576/Cs. | ||||||
15763575
|
Fisherbrand™
02911806A |
24-400 | 4100/Cs. |
N/A
|
N/A | ||||
11598172
|
Fisherbrand™
02912101 |
28-400 | 576/Cs. | ||||||
15512361
|
Fisherbrand™
02912062 |
33-400 | 576/Cs. | ||||||
11538182
|
Fisherbrand™
02911727 |
38-400 | 1600/Cs. | ||||||
15573261
|
Fisherbrand™
02912063 |
38-400 | 576/Cs. | ||||||
Description
PolyCone Lined Phenolic Caps form an exceptionally tight seal and offer a good chemical barrier. This liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off. Great for everything from liquids, to soaps, to macro-invertebrates.
- Liner molds itself around the sealing areas of finish as cap is screwed on
- Eliminates leakage, evaporation, contamination, binding and backing off
Specifications
Black | |
Phenolic Resin | |
PolyCone | |
Polycone Cap |