missing translation for 'onlineSavingsMsg'
Learn More

Bis[2-(dicyclohexylphosphino)ethyl]amine, 97+%, Thermo Scientific Chemicals

Product Code. 15426748
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 550373-32-5 | C28H53NP2 | 465.69 g/mol

It is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Air Sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 550373-32-5
Molecular Formula C28H53NP2
Molecular Weight (g/mol) 465.69
MDL Number MFCD17014019
InChI Key OQJIHPKBLHGKJN-UHFFFAOYSA-N
Synonym bis 2-dicyclohexylphosphino ethyl amine, bis 2-dicyclohexylphosphanyl ethyl amine, 2-dicyclohexylphosphanyl-n-2-dicyclohexylphosphanyl ethyl ethan-1-amine
PubChem CID 58194775
IUPAC Name 2-dicyclohexylphosphanyl-N-(2-dicyclohexylphosphanylethyl)ethanamine
SMILES C(CP(C1CCCCC1)C1CCCCC1)NCCP(C1CCCCC1)C1CCCCC1

Specifications

Melting Point 64°C to 69°C
Linear Formula [(C6H11)2PCH2CH2]2NH
Quantity 1 g
UN Number UN3259
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 465.67
Percent Purity ≥97%
Chemical Name or Material Bis[2-(dicyclohexylphosphino)ethyl]amine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.