Learn More
Agilent BioTek Complete Waste System
Provides supply and waste containment with optimal evacuation efficiency for 384- and 1536-well microplates
10482.00CHF - 11430.00CHF
Product Code | Brand | Includes | For Use With (Equipment) | Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | For Use With (Equipment) | Type | Price | Quantity & Availability | |||
15645437
![]() |
Agilent BioTek
7100751 |
ELx 405 Microplate Washer | Complete Dispense/High Flow Waste System |
10482.00CHF
Each |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||
16522252
![]() |
Agilent BioTek
7100752 |
115V high flow vacuum pump assembly, vacuum set with level sense assembly with 10 L bottles, dispense system complete assembly with 10 L bottles | EL406 Microplate Washer, ELx 405 Microplate Washer | Complete Dispense/High Flow Waste System |
11430.00CHF
Each |
Estimated Shipment: 13-05-2025
Log in to see stock. |
|||
Description
- Includes complete high flow waste system and complete dispense system
- 115V/230V