Learn More
Bel-Art™ Glove Box Economy Sleeve Gloves
One piece, bonded neoprene protection against liquid or gas penetration
3861.92 NOK - 3960.77 NOK
Specifications
Color | Black |
---|---|
Hand | Specific |
Latex Free | No |
Material | Bonded Neoprene |
Product Type | Glove Box Gloves |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
11767385
|
Bel-Art™
H50025-0546 |
9 |
3960.77 NOK
Pack of 2 |
Estimated Shipment: 01-10-2025
Log in to see stock. |
|||||
11757385
|
Bel-Art™
H50025-0544 |
10 |
3861.92 NOK
Pack of 2 |
Estimated Shipment: 26-09-2025
Log in to see stock. |
|||||
Description
- Single piece glove with sleeve
- For glove boxes with 8 in. glove ports
- Requires clamping rings (19-821-983) to hold gloves in place
- Sold in pairs
Specifications
Black | |
No | |
Glove Box Gloves |
Specific | |
Bonded Neoprene |