Learn More
Bel-Art™ 28-Piece Fittings Assortment Kit
Compatible with tubing from 3.2 to 12.7mm. Bel Art™ 28-Piece Fittings Assortment Kit are for the most common lab connections.
Brand:  Bel-Art™ F19626-0000
1043.41 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 11382214
  • 1180.00 NOK / Pack of 28
Only null left
Estimated Shipment: 20-10-2025
Description
  • Molded of autoclavable FDA grade polypropylene except quick disconnect fittings which are polyethylene and not autoclavable or FDA grade
  • Kit contains: 6 Straight Tubing Connectors, 7 Stepped Tubing Connectors, 5 Wye (Y) Tubing Connectors, 3 “T” shaped Tubing Connectors, 5 Quick Disconnects and 2 flow control Plastic Tubing Clamps
  • Plastic organizer box has room for adding other fittings and parts
Specifications
Specifications
Autoclavable
0.19 to 0.5 in. Tubing
Fittings Kit
Polypropylene
Natural