Learn More
BCAR1 Mouse anti-Human, Unconjugated, Clone: 2D11C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67215-1-IG-20UL
This item is not returnable.
View return policy
Description
The BCAR1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BCAR1. This antibody recognizes human antigen. The BCAR1 antibody has been validated for the following applications: FC, IF, IHC, WB, ELISA analysis.
Specifications
BCAR1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, Immunofluorescence, Flow Cytometry, ELISA | |
2D11C10 | |
Unconjugated | |
20 μL | |
9564 | |
IgG2a |