Learn More
Azlon™ Polypropylene Powder Funnels
Constructed of durable, lightweight Polypropylene. Azlon™ Polypropylene Powder Funnel features a parallel stem that minimises 'bridging' of powder to improve flow. The two largest sizes have vertical-sided tops to prevent spillage.
202.23 NOK - 610.00 NOK
Product Code | Brand | Diameter (Metric)Top | Diameter (Metric) Stem | Height (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Diameter (Metric) Stem | Height (Metric) | Price | Quantity & Availability | |||
12571506
|
Azlon
FWC208 |
120mm | 30mm | 106 mm |
480.00 NOK
Pack of 5 |
Estimated Shipment: 26-11-2025
Log in to see stock. |
|||
10589730
|
Azlon
FWC210 |
150mm | 36mm | 135 mm |
610.00 NOK
Pack of 5 |
Estimated Shipment: 18-11-2025
Log in to see stock. |
|||
12030783
|
Azlon
FWC212 |
290mm | 35mm | 208 mm |
585.00 NOK
Each |
Estimated Shipment: 02-12-2025
Log in to see stock. |
|||
12040783
|
Azlon
FWC214 |
420mm | 48mm | 270 mm | |||||
12074628
|
Azlon
FWC202 |
65mm | 18mm | 69 mm | |||||
12561506
|
Azlon
FWC204 |
80mm | 22mm | 74 mm | |||||
12054638
|
Azlon
FWC206 |
100mm | 26mm | 90 mm |
348.00 NOK
Pack of 5 |
Estimated Shipment: 18-11-2025
Log in to see stock. |
|||
Description
Constructed of durable, lightweight Polypropylene. Azlon™ Polypropylene Powder Funnel features a parallel stem that minimises 'bridging' of powder to improve flow. The two largest sizes have vertical-sided tops to prevent spillage.
- Lightweight polypropylene powder funnel
- Unbreakable in normal use
- Parallel stem minimises 'bridging' of powder to improve flow
- The two largest sizes have vertical sided tops
Specifications
Polypropylene | |
Autoclavable |