Learn More
Axygen™ Screw Cap Tubes without Caps: Conical with Conical Bottoms
Conical tubes
Brand:  Axygen™ ST-150-C-S
6957.85 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12719608
  • 7006.55 NOK / Pack of 4000
Only null left
Estimated Shipment: 07-01-2026
Description
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNase, DNase, pyrogen-free
Clear
Bag
Not Included
1.5 mL
Polypropylene
Sterile
Conical