Learn More
Telstar™ AV-100 Vertical Laminar Flow Bench
The AV-100 Vertical Laminar Flow Bench, with 1 HEPA filter for down flow, allows operation in sterile and particle-free conditions due to the continuous flushing of the working area by a unidirectional vertical and ultrafiltered air flow, thus assuring full product protection.
Brand: Telstar™ 31400
This item is not returnable.
View return policy
Description
- Supplied with working area CLASS 10 as defined by U.S. Fed. Std. 209 a B.S. 5295
- Specially designed for handling of non-pathogenic biological samples, cell and tissue cultures, controls in microbiology, pharmaceutical products preparation, electronics, optics and more
- Cabinets made of stove-enamel coated steel
- Modern and accurate design makes it possible to change the filters from inside the cabinet, having reduced external dimensions with suitable working area
- Polished stainless steel working surface divided into three parts for easy removal for sterilizing
- Removable side walls and front window with gas springs, both made of UV resistant tempered glass
- Materials and construction according to international standards, with high quality components and easy service
- Digital control board with LCD display including selection of air speed, button for UV irradiation, indicative LEDS for air speed, lighting, UV clogging and more, alarm LED and air speed adjustment
- ON/OFF selector
- Electrical power socket inside of the cabinet
Specifications
Stove Enamel | |
Stove Enamel-coated Steel, Polished Stainless Steel Work Surface | |
Handling of non-pathogenic biological samples, cell and tissue cultures, controls in microbiology, pharmaceutics, electronics, optics, etc. | |
50 Hz, Single Phase | |
0.6 kW | |
140 kg |
HEPA | |
845 x 1260 x 1290 mm | |
700 mm | |
>800 lx | |
220 V |