Learn More
3M™ Aura™ Particulate Healthcare Respirator FFP2 Type IIR
Particulate healthcare respirator is our foldable, 3-panel respirator with a sculpted nose panel that conforms to a broad range of face shapes.
Brand: 3M™ 7000088720
This item is not returnable.
View return policy
Description
- FFP2 (NPF 12) protection against hazardous airborne contaminates in medical environments
- 3-panel design allows greater facial movement and wide-ranging fit
- Low resistance filter technology helps you breathe easily
- Sculpted upper panel conforms to the nose and contours of the face with a soft foam for an effective seal
- Embossed top panel helps reduce eyewear fogging
- Chin tab designed for easy positioning and adjustment
- Hygienic packaging protects the respirator from contamination before use
- Outer cover provides resistance to fluid splashes
- Approved to the European Medical Device Directive (93/42/EEC) and EN14683:2005 Type IIR for use as a surgical mask
Specifications
White, Blue | |
No | |
No |
General Purpose Particulate Respirator | |
Non-sterile |