Learn More
Analytical Volumetric Sodium Thiosulfate, 0.05N, 0.05M, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Packaging:
1L
Unit Size:
1 L
Specifications
Specifications
Quantity | 1 L |
Concentration or Composition (by Analyte or Components) | 0.05M |
Packaging | 1L |