Learn More
BRAND™ Amber USP Measuring Flask
With glass stopper
Brand:  BRAND™ 957470
1086.23 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15376379
  • 1155.00 NOK / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Description
  • Boro 3.3
  • To DIN EN ISO 1042
  • Calibrated to contain (TC, In)
  • With USP batch certificate
  • TRUSTED_SUSTAINABILITY
    Specifications
    Specifications
    Glass
    500 mL
    Ground Joint NS 19/26
    Amber
    Borosilicate Glass (3.3)
    A