Learn More
BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers, with Certificate
Amber glass to protect light-sensitive contents. Delivered with individual USP conformity certificate.
Brand: BRAND™ 957487
700.17 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Manufactured from Boro 3.3 glass
- Acc. to DIN EN ISO 1042
- Delivered with individual USP conformity certificate
- Class A, DE-M marking
Especificaciones
Polypropylene | |
100 mL | |
1 | |
NS 14/23 | |
A |
13 mm | |
Borosilicate Glass (3.3) | |
Volumetric flask | |
±0.08 mL | |
Stopper |