Learn More
Macherey-Nagel™ Aluminum Oxide, 90 Basic
Brand:  Macherey-Nagel™ 815010.1
Product Code. 10508541
  • 641.00 NOK / 1kg
In Stock
to see stock.
Explore more special offers
Specifications
50 to 200 μm
9.5 ±0.3
1 kg
130 m2/g