Learn More
Rogo Sampaic™ Aluminum Lab Jack
€ 177.00 - € 305.00
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
11747265
|
Rogo Sampaic™
FCD002 |
65 to 280 mm |
€ 177.00
Each |
Estimated Shipment: 30-06-2025
Log in to see stock. |
|||||
11757265
|
Rogo Sampaic™
FCD003 |
65 to 290 mm |
€ 305.00
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
|||||
11737265
|
Rogo Sampaic™
FCD001 |
65 mm |
€ 185.00
Each |
Estimated Shipment: 27-06-2025
Log in to see stock. |
|||||
Specifications
Anodized Aluminum | |
Lab Jack | |
1 Ea. |