Learn More
Cole-Parmer™ Aluminum Gear Flowmeter
Brand:  Cole-Parmer™ 32928-01
Product Code. 11775838
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
Gear Flowmeter
Aluminum
1/4 in. NPT (F)
10 to 24 VDC at 25 mA
85°C
±0.5% (Reading)
3000 psig
Ideal for measuring the flow of oils, grease, fuels, solvents, and other nonabrasive lubricating liquids
±0.1%
Product Title
Cole-Parmer™ Aluminum Gear Flowmeter > Aluminum