Learn More
3M™ E-A-R™ E-A-Rsoft FX Earplugs
Offers high attenuation and are made from a slow expanding, PU foam material. 3M E-A-Rsoft FX Earplug – 39dB.
690.00 NOK - 1630.00 NOK
Specifications
Color | Neon Green |
---|---|
Quantity | 200/Pk. |
Product Type | Ear Plugs |
Style | Roll-down |
Product Code | Brand | Corded | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Corded | Price | Quantity & Availability | |||||
12940325
|
3M™
7000052710 |
Uncorded |
690.00 NOK
200 pairs |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||||
12950325
|
3M™
7000052711 |
Corded |
1630.00 NOK
200 pairs |
Estimated Shipment: 08-09-2025
Log in to see stock. |
|||||
Description
- Highest available attenuation rated earplug at 39dB
- Smooth tapered finish makes fitting easier and helps form a comfortable fit
- Flange shape, when rolled down, serves as a very effective insertion device
- Ideal for those looking for higher attenuation
Specifications
Neon Green | |
200/Pk. | |
Ear Plugs | |
Roll-down |