Learn More
Ansell™ AlphaTec™MICROCHEM™4000 Model 215 Apron with Sleeves
MICROCHEM™ 4000 is designed to provide anexceptional barrier against organic and inorganicchemicals and biological agents
Brand: Ansell™ GR40T-00215-04
11762.22 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Velcro fastening to neck
- Tie fastening at the waist
- Double cuff with knitted
- inner cuff
- Welded and taped seams
Specifications
Apron | |
Multi-layer Barrier Laminate Fabric | |
Limited-use | |
Non-sterile | |
Yes | |
Abrasions, acids, anti-static, bloodborne pathogens, chemicals | |
Alphatec™ 4000, Model 215 |
Large | |
Green | |
Hook and Loop, Waist Ties | |
Double Inner Knit | |
None | |
EN 14605: 2005 +A1:2009, Type PB (3); EN 14605: 2005 +A1:2009, Type PB (4); EN14126: 2003, Types PB(3)-B, PB (4)-B | |
Welded and Taped |