Learn More
Ansell AlphaTec™ 37-210 Reusable Nitrile Gloves with UltraGrip™ Technology
UltraGrip™ Technology delivers an innovative raised diamond grip on the glove's surface for enhanced grip capabilities in wet and oily conditions.
229.00 NOK
Specifications
ANSI Abrasion Rating | ANSI 4 |
---|---|
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009, REACH Compliant |
Color | Blue |
Disposable | Reusable |
Hand | Specific |
Description
- Reusable nitrile gloves designed specifically for food processing and handling
- Raised diamond-pattern enhances grip in wet and oily conditions
- Gloves maintain flexibility and dexterity, even at low temperatures
- Use alone or as outer glove over thermal liners or cut-resistant gloves
- Latex-free formulation presents no risk of Type 1 allergies
Specifications
ANSI 4 | |
Blue | |
Specific | |
Nitrile | |
3001X | |
JKT |
EN 420:2003 + A1:2009, REACH Compliant | |
Reusable | |
Yes | |
8 mil | |
Type B | |
37-210 |