Learn More
Thermo Scientific™ Grade 25 Sapphire Spheres, Dia.: 2.5mm
649.00 NOK - 2418.00 NOK
Specifications
Product Type Sapphire Sphere
Diameter (Metric) 2.5 mm
Grade Grade 25
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15481757
View Documents
Thermo Scientific™
042062.KJ
5 Pieces This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11336009
View Documents
Thermo Scientific™
042062.KM
25 Pieces This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Sapphire Sphere
2.5 mm
Grade 25