Learn More
ALDH1L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17390-1-AP-20UL
This item is not returnable.
View return policy
Description
ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.Specifications
ALDH1L1 | |
Polyclonal | |
Unconjugated | |
ALDH1L1 | |
10 FTHFDH, ALDH1L1, FTHFD | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107747, 10840, 64392 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75891, P28037, Q8R0Y6 | |
Aldh1l1 | |
ALDH1L1 Fusion Protein Ag11285 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |