Learn More
Al-24 Tube, Both ends open, O.D.: 50mm, I.D.: 40mm, Thermo Scientific™
848.23 NOK - 7830.00 NOK
Specifications
Chemical Name or Material Al-24 Tube
Name Note Both ends open
View More Specs
Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394127
View Documents
Thermo Scientific Alfa Aesar
031608.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11354127
View Documents
Thermo Scientific Alfa Aesar
031608.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11364127
View Documents
Thermo Scientific Alfa Aesar
031608.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11344127
View Documents
Thermo Scientific Alfa Aesar
031608.EH
600 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11374127
View Documents
Thermo Scientific Alfa Aesar
031608.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11384127
View Documents
Thermo Scientific Alfa Aesar
031608.EO
1000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Al-24 Tube
Both ends open
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures