Learn More
Al-23 Tube, Both ends open (Thin walls), O.D.: 20mm, I.D.: 19mm, Thermo Scientific™
746.00 NOK - 1692.30 NOK
Specifications
Chemical Name or Material Al-23 Tube
Name Note Both ends open (Thin walls)
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11358267
View Documents
Thermo Scientific Alfa Aesar
031712.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11348267
View Documents
Thermo Scientific Alfa Aesar
031712.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11328267
View Documents
Thermo Scientific Alfa Aesar
031712.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Al-23 Tube
Both ends open (Thin walls)
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures