Learn More
Al-23 Solid Round Rod, Dia.: 0.8mm, Thermo Scientific™
166.12 NOK - 446.44 NOK
Specifications
Chemical Name or Material Al-23 Solid Round Rod
View More Specs
Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11353038
View Documents
Thermo Scientific Alfa Aesar
032922.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11383038
View Documents
Thermo Scientific Alfa Aesar
032922.DR
200 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11363038
View Documents
Thermo Scientific Alfa Aesar
032922.DT
250 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11373038
View Documents
Thermo Scientific Alfa Aesar
032922.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Al-23 Solid Round Rod
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures