Learn More
Al-23 Solid Rod, Round, Dia.: 6mm, Thermo Scientific Chemicals
316.66 NOK - 1960.00 NOK
Specifications
Chemical Name or Material Al-23 Solid Rod
Form Rod
View More Specs
Products 7
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11343088
View Documents
Thermo Scientific Alfa Aesar
032933.DM
100 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11383088
View Documents
Thermo Scientific Alfa Aesar
032933.DR
200 mm
794.00 NOK
200mm
11333088
View Documents
Thermo Scientific Alfa Aesar
032933.DT
250 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11313088
View Documents
Thermo Scientific Alfa Aesar
032933.DW
300 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11363088
View Documents
Thermo Scientific Alfa Aesar
032933.EH
600 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11373088
View Documents
Thermo Scientific Alfa Aesar
032933.EM
800 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11353088
View Documents
Thermo Scientific Alfa Aesar
032933.EO
1000 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Al-23 Solid Rod
Rod
Videos
Safety and Handling
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures