Learn More
Al-23 Protective Tube, One End Closed, O.D.: 8mm, I.D.: 5mm, Wall.: 1.5mm, Thermo Scientific™
239.83 NOK - 4368.00 NOK
Specifications
Chemical Name or Material | Al-23 Protective Tube |
---|---|
Name Note | One end closed |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11322668
|
Thermo Scientific Alfa Aesar
030044.DR |
200 mm | N/A | ||||||
11352668
|
Thermo Scientific Alfa Aesar
030044.DU |
270 mm | N/A | ||||||
11342668
|
Thermo Scientific Alfa Aesar
030044.DY |
375 mm | N/A | ||||||
11382668
|
Thermo Scientific Alfa Aesar
030044.EE |
530 mm | N/A | ||||||
11312668
|
Thermo Scientific Alfa Aesar
030044.EK |
740 mm | N/A | ||||||
11362668
|
Thermo Scientific Alfa Aesar
030044.EQ |
1030 mm | N/A | ||||||
11372668
|
Thermo Scientific Alfa Aesar
030044.EV |
1430 mm | N/A | ||||||
11332668
|
Thermo Scientific Alfa Aesar
030044.FA |
2030 mm | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Al-23 Protective Tube | |
One end closed |
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures