Learn More
Al-23 Insulating Tube, Both Ends Open (Thin Wall), O.D.: 2mm, I.D.: 1.2mm, Thermo Scientific™
Click to view available options
Quantity:
100 mm
200 mm
250 mm
300 mm
Unit Size:
100mm
200mm
250mm
300mm
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Chemical Name or Material | Al-23 Insulating Tube |
Name Note | Both ends open (Thin Wall) |
Quantity | 200 mm |
Form | Tube |