Learn More
Al-23 Insulating Tube, Both Ends Open (Thin Wall), O.D.: 0.8mm, I.D.: 0.4mm, Thermo Scientific™
231.52 NOK - 414.25 NOK
Specifications
Chemical Name or Material | Al-23 Insulating Tube |
---|---|
Name Note | Both ends open (Thin Wall) |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Al-23 Insulating Tube | |
Both ends open (Thin Wall) |
Safety and Handling
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures