Learn More
Memmert™ Additional Stainless Steel Grid for UF450, Reinforced Chamber Option
Additional grid features a sturdy stainless steel construction.
Brand: Memmert™ UF450B32190
6715.10 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Reinforced
- Stainless Steel
- Electropoloished
- Max. loading 60 kg
Specifications
UF450 Oven | |
Reinforced Grid | |
Stainless Steel | |
60 kg |