Learn More
Eppendorf™ Additional Humidity Tray
For Use With: Innova CO-14 Incubator
Brand: Eppendorf™ P0628-6200
1960.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Additional Humidity Tray
Specifications
0.3 L | |
Innova CO-14 Incubator | |
Tray |