Learn More
Eppendorf™ Adapter, for 7 blood collection tubes 9 mL, for Rotor FA-6x250, 2 pcs.
Brand:  Eppendorf™ 5920764004
3870.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 16252411
  • 3885.48 NOK / Pack of 2
Description
Adapter, for 7 blood collection tubes 9 mL, for Rotor FA-6x250, 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapters, Buckets, and Accessories
Rotor FA-6x250
7
2