Learn More
Eppendorf™ Adapter for Rotor F-35-6-30
Designed for use with Eppendorf Rotors and Centrifuges
1255.00 NOK
Specifications
Type Adapter
No. of Places 12
View More Specs
Products 4
Product Code Brand For Use With (Equipment) Max. Tube Length Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Max. Tube Length Price Quantity & Availability  
13216589
View Documents
Eppendorf™
5427742000
for Rotor F-35-6-30, large rotor bore 65 to 89 mm
1255.00 NOK
Pack of 2
View alternative products
Save Up to 
»
13276579
View Documents
Eppendorf™
5427741003
for Rotor F-35-6-30, small rotor bore 90 to 110 mm
1255.00 NOK
Pack of 2
View alternative products
Save Up to 
»
13286579
View Documents
Eppendorf™
5427740007
for Rotor F-35-6-30, small rotor bore 65 to 89 mm
1255.00 NOK
Pack of 2
View alternative products
Save Up to 
»
13256868
View Documents
Eppendorf™
5427743006
for Rotor F-35-6-30, large rotor bore 90 to 110 mm
1255.00 NOK
Pack of 2
View alternative products
Save Up to 
»
Description
Description
  • For Rotor F-35-6-30
Specifications
Specifications
Adapter
12
Documents
Product Certifications
Product Content Correction
Product Title
Eppendorf™ Adapter for Rotor F-35-6-30