Learn More
Eppendorf™ Adapter, for 12 tubes 30 mL, for Rotor S-4xUniversal-Large, 2 pcs.
Brand:  Eppendorf™ 5920743007
4070.00 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 16030772
  • 4086.28 NOK / Each
Description
Adapter, for 12 tubes 30 mL, for Rotor S-4xUniversal-Large, 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapters, Buckets, and Accessories
Rotor S-4 xUniversal-Large
12
2