Learn More
Acetic acid, Reagent Grade, ≥99.0%, Honeywell™
Product Code. 15665580 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
5 L
Packaging:
Poly Bottle
Reagent Grade,≥99.0%
  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 64-19-7
Molecular Formula C2H4O2
Molecular Weight (g/mol) 60.05
MDL Number MFCD00036152
InChI Key QTBSBXVTEAMEQO-UHFFFAOYSA-N
Synonym ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure
PubChem CID 176
ChEBI CHEBI:15366
IUPAC Name acetic acid
SMILES CC(O)=O
Specifications
Melting Point 16.2°C (literature)
Boiling Point 118°C
CAS Max % 100.0000%
Assay Percent Range 99.0 to 100.5%
Linear Formula CH3CO2H
UN Number UN2789
Beilstein 506007
Formula Weight 60.05
Grade Reagent
Density 1.049 g/mL at 25 °C(lit.)
Quantity 2.5 L
Flash Point 40°C
Packaging Poly Bottle
Chemical Name or Material Acetic acid, Reagent Grade, ≥99.0%