Learn More
Acetic Acid 1M, Chem Lab™
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Specifications
Specifications
pH | <3 |
Color | Colorless |
Solubility Information | Soluble in water |
Density | 1g/mL |
Quantity | 1 L |
Odor | Specific |
Physical Form | Liquid |