Learn More
Sartorius Accessories for Vivacell Models
Accessories for various Vivacell units
942.59 NOK - 1465.00 NOK
Specifications
Disposable | Reusable |
---|---|
Filter Type | Ultrafiltration |
Sterility | Non-sterile |
Type | Accessory for Vivacell Model |
Product Code | Brand | For Use With (Equipment) | For Use With (Application) | Includes | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | For Use With (Application) | Includes | Price | Quantity & Availability | |||
10663382
|
Sartorius
VCA005 |
For Vivacell 20 and 70; For Pressure-fuge head | Charge Valve | Not Applicable |
1465.00 NOK
Each |
In Stock
Log in to see stock. |
|||
10059730
|
Sartorius
VCA009 |
Vivacell 250 | Maintenance Kit | Sample Reservoir, Filtrate Container, O-ring Seals | N/A | ||||
Description
- Use charge value with pressure-fuge head
- Maintenance Kit includes sample reservoir, filtrate container and 'O' ring seals
Specifications
Reusable | |
Ultrafiltration | |
Non-sterile | |
Accessory for Vivacell Model |